Learn More
Proteintech HumanKine™ recombinant human EGF protein
HumanKine™ recombinant protein
Brand: Proteintech HZ-1326-1000UG
Specifications
P01133 | |
1950 | |
100 μg | |
Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.) | |
Human | |
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 xPBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA). | |
Lyophilized |
1 x PBS, See Certificate of Analysis for details | |
7 kDa, monomer, glycosylated | |
HEK293 | |
<1.0 EU/μg | |
Recombinant | |
0.1-0.6 ng/mL EC50 | |
>95% |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.