Learn More
Cytiva HyClone™ FetalClone™ I Serum (U.S.)
Optimized for growth of hybridomas, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ I Serum, a fetal bovine serum (FBS) alternative containing comparable IgG levels to those found in FBS.
Brand: Cytiva SH30080.03
193.42 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : A0
Additional Details : Weight : 0.45000kg
Description
Specifications
Albumin, Iron, Blood Urea Nitrogen, Osmolality, Percent Iron Saturation, Creatinine, Gamma Globulin, Glucose, SGPT, Sodium, TIBC, Alkaline Phosphatase, Total Bilirubin, Total Protein, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
United States | |
500 mL | |
Sequential-filtered, 100 nm | |
≤ 10 EU/mL | |
≤ 20 mg/dL | |
No |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
FetalClone I Serum (U.S.) | |
FBS Replacement Serum | |
Sterile | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
No | |
Typically less than 200 μg/mL | |
USDA Inspected |