missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ FetalClone™ III Serum (U.S.)

Designed for fibroblast applications, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ III Serum, the most widely applicable fetal bovine serum (FBS) alternative.

Brand:  Cytiva SH30109.03IR25-40

 View more versions of this product

Product Code. 16281102

  • £372.00 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Economical alternative to FBS
  • Performance comparable with FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Total Protein,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Osmolality,Iron,TIBC,Percent Iron Saturation,Glucose,IgG
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
FBS Replacement Serum
Sterile
Irradiated (25 to 40 doses)
≤ 10 EU/mL
≤ 20 mg/dL
HyClone™
USDA Inspected
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
United States
500 mL
Sequential-filtered, 100 nm
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
No
Typically less than 2 μg/mL
No
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Hazard Statement
  • EUH210-Safety data sheet available on request.

Supplemental information
  • UNKNOWN ACUTE-9.9 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 9.9% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva HyClone™ FetalClone™ III Serum (U.S.) > Irradiated (25 to 40 doses); 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.