missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ US Bovine Calf Serum

A cost effective alternative to Fetal Bovine Serum (FBS). Cytiva HyClone™ US Bovine Calf Serum is defined, irradiated, heat inactivated serum of US origin that is filtered through three sequential 0.1μm pore size-rated filters.

Brand:  Cytiva SH30073.03HI

Product Code. 12389812

  • £77.85 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

HyClone Bovine Calf Serum, U.S. origin is collected by venipuncture from formula-fed veal calves, which produce exceptionally high levels of transferrin. Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.

  • Sourced in the USA with complete traceability back to original source
  • Cost-effective alternative to FBS
  • Age of animal at time of collection approximately 16-24 weeks
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Insulin Gamma Globulin Alkaline Phosphatase Lactate Dehydrogenase SGPT SGOT pH Albumin Blood Urea Nitrogen Creatinine Total Bilirubin Sodium Potassium Calcium Chloride Inorganic Phosphorus Iron TIBC Percent Saturation Glucose IgG
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
16 to 24 weeks
United States
500 mL
Sequential-filtered, 100 nm
Yes
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Heat Inactivated; 500 mL
Bovine
Calf Bovine Serum
Sterile
Bluetongue Bovine Adenovirus Bovine Parvovirus Bovine RSV BVDV Cytopathogenic Agents Hemadsorbing Agents Rabies Virus Reovirus
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva HyClone™ US Bovine Calf Serum > Quantity: 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.