missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Medium K, Honeywell Fluka™

Medium for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free).

Brand:  Honeywell Fluka 34698-1L

91.72 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.



 View more versions of this product

Product Code. 15604360

  • £96.55 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Advantages of Hydranal one-component reagents:

  • Unlimited water capacity
  • Convenient and easy to use
  • The greatest flexibility in working media selection
  • reduced toxicity compared to methanol based systems
  • Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
  • Long shelf life (three years)

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HYDRANAL™-Medium K
56°C (132.8°F)
101023-55-6, 75-89-8, 107-98-2, 67-66-3
<5.00, <20.00, <30.00, <60.00
Polycoated bottle
One component volumetry
1.25g/mL
1 L
≥1.00, ≥10.00, ≥20.00, ≥30.00
58 °C
Liquid
UN1992
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    18

    Hours

    17

    Minutes

    41

    Seconds

    35
  • Multiple Manufacturers
    Explore our new promotion and get up to 70% off some of our most popular products.
    Promo Code: 24127

    Expires2025-05-31

    Days

    49

    Hours

    17

    Minutes

    41

    Seconds

    35
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
HYDRANAL™ - Medium K, Honeywell Fluka™ > 1L, Polycoated Bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.