Learn More
Thermo Scientific™ HyperSep™ Retain PEP Cartridges
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Cartridges.
Brand: Thermo Scientific™ 60107-218
396.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Cartridges, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Features:Ideal for large samples, Hypersep Retain cartridges conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
- High, consistent recoveries
- Fast and easy sample preparation and method development
- Reproducible, reliable sample preparation in a traditional format
- Available in a wide range of chemistries and bed weights
- Drugs and metabolites in biological fluids
- Peptides in serum, plasma or biological fluids
- Environmental samples

Specifications
6 mL | |
1000 mg | |
Thermo Scientific HyperSep-96 System | |
HyperSep | |
Solid Phase Extraction Cartridge | |
Pack of 30 |
Retain PEP 1 g/6 mL Cartridge pk30 | |
Retention of Polar and Nonpolar Analytes | |
40 to 60 μm | |
HyperSep Urea-Modified Polystyrene DVB | |
6 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.