missing translation for 'onlineSavingsMsg'
Learn More
Iminostilbene, 97%, Thermo Scientific Chemicals Product Code.: 11469237

Iminostilbene, 97%, Thermo Scientific Chemicals

Product Code. 11469237
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11469237

Brand: Thermo Scientific Alfa Aesar B21323.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Iminostilbene has good antioxidant effects. N-acetyl iminostilbene was synthesized by incubating iminostilbene (2.0020 g, 10.360 mmol) with acetic anhydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iminostilbene has good antioxidant effects. N-acetyl iminostilbene was synthesized by incubating iminostilbene (2.0020 g, 10.360 mmol) with acetic anhydride.

Solubility
Soluble in ethyl acetate (25 mg/ml), water (partly), dioxane, chloroform, and DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 256-96-2
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD00005071
InChI Key LCGTWRLJTMHIQZ-UHFFFAOYSA-N
Synonym iminostilbene, 5h-dibenzo b,f azepine, 5h-dibenz b,f azepine, 2,2'-iminostilbene, dibenz b,f azepine, 2,3,6,7-dibenzazepine, stilbene, 2,2'-iminoShow MoreShow Less
PubChem CID 9212
ChEBI CHEBI:47802
IUPAC Name 11H-benzo[b][1]benzazepine
SMILES C1=CC=C2C(=C1)C=CC3=CC=CC=C3N2

Specifications

Melting Point 197°C to 199°C
Quantity 100 g
Beilstein 1343358
Solubility Information Soluble in ethyl acetate (25mg/ml),water (partly),dioxane,chloroform,and DMSO.
Formula Weight 193.25
Percent Purity 97%
Chemical Name or Material Iminostilbene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10214444 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberB21323.22,B21323.14,B21323.36
Lot NumberG2088C Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21323.22,B21323.14,B21323.36
Lot Number10214444 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21323.22,B21323.14,B21323.36
Lot Number10230640 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21323.22,B21323.14,B21323.36
Lot NumberE3883A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21323.22,B21323.14,B21323.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iminostilbene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.