Learn More
METTLER TOLEDO™ InLab™ Easy pH Electrodes
A durable, polysulfone pH electrode with an integral bulb protector and gel electrolyte to avoid the inconvenience of refilling in field. Temperature sensors sold separately.
Brand: METTLER TOLEDO™ 51343011
139.05 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Aquarium water
- Soft drinks
- Beer
- General lab
Accessories & Care
- Ceramic junction
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
- Temperature sensors sold separately (NTC 30 kΩ: 51300164, PT1000: 51300165)

Specifications
BNC | |
Polysulfone | |
0°C to 80°C | |
Gel | |
Fixed Cable to sensor | |
U Glass | |
NTC 30 kilohm | |
3mol/L KCl | |
0°C to 80°C | |
120 mm | |
0.47 in. |
Ceramic | |
0 to 14 | |
Combination Electrode | |
Low maintenance, field measurement and educational use | |
Ag/AgCl | |
InLab™ Easy BNC | |
<20 sec. | |
32°F to 176°F | |
4.72 in. | |
1.2 m | |
12 mm |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
13Hours
6Minutes
57Seconds
2 - Multiple Manufacturers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
88Hours
6Minutes
57Seconds
2New Offer Save Up to 85%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.