Learn More
Invitrogen™ S.c. EasyComp™ Transformation Kit
Designed for rapid preparation of transformation-competent Saccharomyces cerevisiae cells
Brand: Invitrogen™ K505001
430.10 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Includes: The S.c. EasyComp Transformation Kit provides enough reagents to prepare 6 batches of competent cells. Each batch is sufficient for 20 transformations.
Description
- Cells can be made competent in less than 30 minutes
- Competent cell preparation and transformation solutions are optimized for use with S. Cerevisiae
- Provided ready-to-use and tested to ensure successful yeast transformation
- Competent cells can be frozen for future use
Cloning, Protein Expression, Proteins, Expression, Isolation and Analysis, S. cerevisiae Expression Systems, Transformation, Yeast Expression
Specifications
Transformation Kit | |
S. cerevisiae | |
EasyComp™ |
The S.c. EasyComp™ Transformation Kit provides enough reagents to prepare 6 batches of competent cells. Each batch is sufficient for 20 transformations. The kit should be stored at +4°C. All components are guaranteed stable for 6 months when properly stored. | |
1 kit |