Learn More
Invitrogen™ UltraPure™ TBE Buffer, 10X

Compatible with agarose gels
Brand: Invitrogen™ 15581044
17.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
UltraPure 10X TBE Buffer is a sterile-filtered solution of 1M Tris, 0.9 M boric acid, and 0.01M EDTA used to prepare 1X buffer for polyacrylamide and agarose gel electrophoresis.
- UltraPure 10X TBE Buffer is available in 1 L plastic bottle/10 L Cubitainer™ box
- Performance and quality testing: No DNase, RNase, or protease activity detected. Sustainable packaging
Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting
Order Info
Shipping Condition: Room Temperature

Specifications
Running Buffer | |
Contents: One bottle (1 L) UltraPure™ 10X TBE Buffer Storage: Room Temperature |
|
Polyacrylamide Gels, Agarose Gels | |
UltraPure™ | |
1 L |
10 X | |
Bottle | |
Nucleic Acid Gel Electrophoresis, Blotting | |
TBE Buffer | |
Room Temperature |
Safety and Handling
- TBE BUFFER, 10X
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1
Hazard Statement
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : boric acid
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2454557 | Certificate of Analysis | 29/01/2025 | 15581044 |
2703616 | Certificate of Analysis | 29/01/2025 | 15581044 |
2698745 | Certificate of Analysis | 29/01/2025 | 15581044 |
2750330 | Certificate of Analysis | 29/01/2025 | 15581044 |
2744700 | Certificate of Analysis | 29/01/2025 | 15581044 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.