Learn More
Ambion™ iPrep™ Card: Total RNA
iPrep™ Card: Total RNA
Brand:  Ambion™ IS10014
Product Code. 10419793
  • £676.00 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
The iPrep™ Protocol Card is a flash, 512 KB memory card. Each iPrep™ Protocol card is pre-programmed with the purification protocol that directs the volume of reagents used and incubation time.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at room temperature.
iPrep™
Room Temperature
iPrep™
1 Card
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ambion™ iPrep™ Card: Total RNA > 1 card