Learn More
Micronova™ Hazard Bags
Low density, LDPE hazard bags are designed for containing biomedical and hazardous waste in aseptic and controlled environments.
Brand: Micronova™ BL2-32YEIRBB
415.12 GBP valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Description
Micronova Hazard Bags are constructed from FDA compliant, low density, red or yellow polyethylene and can be used with the BLSSH series Micronova BinLiner Stands. The Biohazard bags are designed for containment and disposal of biomedical waste and the Hazardous Material bags are designed for segregation and disposal of hazardous waste
- Low density polyethylene Hazard bags
- Red or Yellow Biohazard and Hazardous Material bags
- Biohazard bags are clearly marked with a Biohazard symbol in black ink
- Hazardous material bags are labeled for waste segregation in the US
- The 32YE liners work with the BLSSH-1515 BinLiner Stand
- The 24YE and 25RE work with the BLSSH-168 BinLiner Stand
- Smaller bags with a star bottom are ideal for round containers
- Larger bags are flat sealed
- Good compatibility with common cleaners
- Gamma irradiated liners are sterile for aseptic areas
Specifications
114 L | |
Five folded liners per double bag | |
20.4 in. | |
Yellow | |
91.4 cm | |
50/Cs. | |
Sterile | |
Transporting Biomedical Waste (Non-Sharps) | |
0.050 mm | |
LDPE | |
15°C to 30°C |
Certificate of Conformance and Certificate of Irradiation Processing; ISO 9001 Certified Facility Meets ASTM D170910-6 Sterility Assurance Level In-Compliance with AAMI Guidelines for Sterilization. | |
51.8 cm | |
30 gal. | |
Single-use | |
81.3 cm | |
STAR: Durable pleated bottom to fit round containers. | |
Not Autoclavable | |
BLSSH series BinStand or similar container | |
36 in. | |
Biohazard Waste Bags |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
047563-000 | Certificate of Quality | 10/02/2025 | BL2-32YEIRBB |
046916-000 | Certificate of Quality | 04/12/2024 | BL2-32YEIRBB |
045913-000 | Certificate of Quality | 23/10/2024 | BL2-32YEIRBB |
044881-000 | Certificate of Quality | 30/08/2024 | BL2-32YEIRBB |
043149-000 | Certificate of Quality | 17/06/2024 | BL2-32YEIRBB |
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
Your input is important to us. Please complete this form to provide feedback related to the content on this product.