Learn More
Integra Biosciences™ PipetBoy Pro Pipettors
Use this pipette controller for precise and efficient serological pipetting. Integra Biosciences™ PipetBoy™ Pro Pipettors feature integrated LED providing an optimal illumination of the area around the pipette for accurate pipetting and preventing eyestrain.
Brand: Integra Biosciences™ 156401
Description
- Accurately displace liquid in the pipette by applying variable finger pressure on the pipettor triggers
- Light as a feather, 190 grams
- Five colors available
- Speed can be set continuously via the thumb wheel
- Red LED indicates when the battery needs to be charged
- Ergonomic design, following the recommendations of hand shape ergonomics
Specifications
1 to 100mL Glass and Plastic Pipette | |
12 mL/sec. | |
Pipette |
Pink | |
PIPETBOY pro | |
190 g |