Learn More
Isopropylidene malonate, 97%, Thermo Scientific Chemicals
Description
Isopropylidene malonate is an important reagent in synthetic chemistry and involved in the formation of new C-C bonds, rings, amides, esters, and acids. The presence of carbonyl groups at C-4 and C-6 positions favors the alkylation and acylation reactions due to its acidic nature. It is also employed as an antimicrobial agent. It is used in the preparation of 4-pyridyl-substituted heterocycles, 2-substituted indoles, isofraxidin and macrocyclic beta-keto lactones. Further, it is involved in the Knoevenagel condensation reaction with aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Isopropylidene malonate is an important reagent in synthetic chemistry and involved in the formation of new C-C bonds, rings, amides, esters, and acids. The presence of carbonyl groups at C-4 and C-6 positions favors the alkylation and acylation reactions due to its acidic nature. It is also employed as an antimicrobial agent. It is used in the preparation of 4-pyridyl-substituted heterocycles, 2-substituted indoles, isofraxidin and macrocyclic beta-keto lactones. Further, it is involved in the Knoevenagel condensation reaction with aldehydes.
Solubility
Soluble in water. Slightly soluble in ethanol and chloroform.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 92°C to 96°C |
Quantity | 100 g |
Beilstein | 117310 |
Sensitivity | Moisture sensitive |
Merck Index | 14,5817 |
Solubility Information | Soluble in water. Slightly soluble in ethanol and chloroform. |
Formula Weight | 144.13 |
Percent Purity | 98% |
Chemical Name or Material | Isopropylidene malonate |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10251564 | Certificate of Analysis | 30/08/2024 | A15603.14,A15603.36,A15603.22 |
10244015 | Certificate of Analysis | 13/10/2022 | A15603.14,A15603.36,A15603.22 |
10153458 | Certificate of Analysis | 23/11/2021 | A15603.14,A15603.36,A15603.22 |
H4077A | Certificate of Analysis | 23/11/2021 | A15603.14,A15603.36,A15603.22 |
10211443 | Certificate of Analysis | 23/11/2021 | A15603.14,A15603.36,A15603.22 |
Safety and Handling


- Isopropylidene malonate
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : Isopropylidene malonate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.