Learn More
Justrite™ Elite Smokers Cease-Fire Cigarette Butt Receptacles Accessories
For safe containment and disposal of cigarette butts
Brand: Justrite™ 26827
Description
- Anchoring Cable: 91.4cm (36 in.) wire rope cable provides added security or greater stability in severe weather
- Brass Padlock: 3/16 in. dia. shackle
- Bucket Liners: special, burn-resistant laminated aluminum/poly construction; include twist-ties; fit any bucket 29.8cm (11.75 in.) wide x 25.4cm (10 in.) high maximum size
Specifications
Silver | |
45.7 cm | |
2 lb. | |
50.8 cm | |
Justrite™ Elite Smokers Cease-Fire Cigarette Butt Receptacles (268503) | |
18 in. |
18 x 20 in. (45.7 x 50.8 cm) | |
10 Pk. | |
0.9 kg | |
Collecting cigarette butts | |
20 in. | |
Disposal Bucket Liner |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.