missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Keratinocyte-SFM Supplement: Human Recombinant Epidermal Growth Factor (EGF 1-53) & Bovine Pituitary Extract (BPE)
Keratinocyte-SFM Supplement: Human Recombinant Epidermal Growth Factor (EGF 1-53) & Bovine Pituitary Extract (BPE)
Brand: Gibco™ 37000015
55.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Human recombinant Epidermal Growth Factor (EGF 1-53) and Bovine Pituitary Extract (BPE).
Specifications
–5 to –30°C | |
Epithelial cells, Keratinocytes | |
Mammalian Cell Culture | |
For research use only. Not for use in diagnostic procedures |
Keratinocyte-SFM Supplement, 1mL, Epithelial Cells, Keratinocytes Cell Type, Human Species, Dry Ice Shipping Condition, Serum-Free Level, Phenol Red Indicator, For Mammalian Cell Culture, –5 to –30°C Storage Temperature | |
Keratinocyte-SFM Supplement | |
1 mL |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Keratinocyte-SFM Supplement: Human Recombinant Epidermal Growth Factor (EGF 1-53) & Bovine Pituitary Extract (BPE) > 1mL
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction