Learn More
Unity Lab Services™ KI Test for Biosafety Cabinet (≤ 1.5 m2)
European KI test for biosafety cabinets 1.5 mt or narrower; Equipment is required to pass certification prior to this service; Required for UK. This is add-on to STARTBSC[PF]
Brand:  Unity Lab Services™ BSCKI15S
Product Code. 17879602
  • £188.24 / Each
Explore more special offers
Alert:
Strongly recommended (and sometimes required) in addition to basic certification (part no BSCCERT)
Description
Description
  • Equipment is required to pass EN12469 certification prior to this test
  • Required for UK
  • This is add-on to STARTBSC[PF] for new equipment, and an add on to BSCCERT for existing equipment
  • Includes travel and labor unless extensive travel is needed
  • All additional works, as i.e. waiting time or correctional works, will be invoiced additionally according to our recent valid service pricelist
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Biosafety Cabinets
1 Day
Thermo Scientific™
BS EN 12469 (Europe) or NSF 49 (US)
European KI Test
Performance Test
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Unity Lab Services™ KI Test for Biosafety Cabinet (≤ 1.5 m2) > Performance Test