Learn More
RETSCH Stainless Steel Grinding Balls
Designed for use with Retsch™ mixer mills. Retsch™ Stainless Steel Grinding Balls are ideal for disruption of biological cells as well as for DNA/RNA extraction.
Brand:  RETSCH 05.368.0034
Code : 50
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 10539711
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • For use with MM 200 and MM 400 Mixer Mills
Specifications
Specifications
Grinding Balls
5 mm
MM 200 and MM 301
Stainless Steel