Learn More
DWK Life Sciences Kimble™ Closures for Sample Vials
Black phenolic closure with PTFE-lined cap
Brand: DWK Life Sciences 73802-24400
This item is not returnable.
View return policy
Description
Black phenolic closure with PTFE-lined cap
- Works with media bottles and screw-thread vials
- Special black phenolic material (wood filled) very resistant to the effects of temperature and steam encountered in autoclaving
- Cemented-in PTFE-faced rubber liner is provided
Specifications
24-400 | |
Screw Cap | |
Phenolic | |
Sample Vials |
PTFE, Rubber | |
Autoclavable | |
Black |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
KIM001 Kimble Chase 08 09 Catalog
File size: 57.1 MB
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Closures for Sample Vials > GPI Thread: 24-400
Spot an opportunity for improvement?Share a Content Correction