missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Arginine, 98+% Product Code.: 11412388

Thermo Scientific Chemicals L-Arginine, 98+%

Product Code. 11412388
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11412388

Brand: Thermo Scientific Chemicals A15738.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Arginine is used for heart and blood vessel conditions which includes congestive heart failure (CHF), chest pain, high blood pressure and coronary artery disease. It plays a vital role in the treatment of cardiovascular disease due to it being antiatherogenic, anti-ischemic, antiplatelet and antithrombotic. It acts as a growth stimulant and is involved in the treatment of erectile dysfunction in men. It is an important ingredient of tooth paste which provides effective relief for sensitive teeth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water. Slightly soluble in ethanol. Insoluble in ethyl ether

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-79-3
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00002635
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
Synonym l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
PubChem CID 6322
ChEBI CHEBI:16467
SMILES NC(CCCN=C(N)N)C(O)=O

Specifications

Melting Point 226°C (decomposition)
Assay Percent Range 98+%
Beilstein 1725413
Merck Index 14,780
Solubility Information Soluble in water. Slightly soluble in ethanol. Insoluble in ethyl ether
IUPAC Name 2-amino-5-[(diaminomethylidene)amino]pentanoic acid
Quantity 500 g
Formula Weight 174.2
Sensitivity Air Sensitive
Percent Purity ≥98%
Chemical Name or Material L-Arginine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0457934 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog NumberA15738.22,A15738.0E,A15738.14,A15738.36
Lot NumberA0450359 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberA15738.22,A15738.0E,A15738.14,A15738.36
Lot NumberN14J069 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA15738.22,A15738.0E,A15738.14,A15738.36
Lot NumberS06J008 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA15738.22,A15738.0E,A15738.14,A15738.36
Lot NumberN14J067 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA15738.22,A15738.0E,A15738.14,A15738.36
compliance-icons
Product Identifier
  • L-Arginine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : L-arginine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Arginine, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.