Learn More
Epredia™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (10X)
Improve the reactivity of many antibodies in formalin-fixed tissues with the Epredia™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (10X).
Brand: Epredia™ AP-9003-500
Description
- Citrate buffer, pH 6.0
- 10X stock solution
- Diluted 10-fold with distilled water before use
- Store at room temperature.
- Stable for up to 18 months.
- Contains no preservatives.
- For periods longer than 3 months, store at 2-8°C.

Specifications
Room temperature | |
Citrate Buffer | |
Immunohistochemistry |
10X | |
500mL | |
6 |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
74384415 | Certificate of Analysis | 13/01/2022 | AP-9003-500 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Reserach Use only