Learn More
Thermo Scientific™ Legionella Latex Test
Rapidly confirm predominant Legionella serogroups grown on selective media by latex agglutination.
Brand: Thermo Scientific™ DR0801M
233.33 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Rapidly differentiate and confirm the identity of predominant Legionella serogroups grown on selective media by latex agglutination using Thermo Scientific™ Legionella Latex Test.
Use this latex agglutination test for the identification of predominant Legionella species grown on plated media from patients with suspected Legionellosis or from environmental sources. The test allows separate identification of Legionella pneumophila serogroup 1 and serogroups 2-14 and detection of seven other Legionella species which have been implicated in human disease.Blue latex particles sensitised with antibody agglutinate in the presence of specific Legionella cell wall antigens to form visible clumps.
The following products are available for use with the Legionella Latex Test and are sold separately:
Legionella Pneumophila Serogroup 1 Test Reagent (Part No. DR0801M)
Legionella Pneumophila Serogroup 2-14 Test Reagent (Part No. DR0802M)
Legionella Species Test Reagent (Part No. DR0800M)—Legionella longbeachae 1 & 2, Legionella bozemanii 1 & 2, Legionella dumoffii, Legionella gormanii, Legionella jordanis, Legionella micdadei and Legionella anisa
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
2°C to 8°C | |
Legionella | |
Legionella Latex Test Kit |
Legionella Pneumophila Serogroup 1 Test Reagent | |
Environmental Source | |
50 Tests/Kit |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6191491 | Certificate of Analysis | 03/02/2025 | DR0801M |
6143610 | Certificate of Analysis | 05/12/2024 | DR0801M |
3761211 | Certificate of Analysis | 07/02/2024 | DR0801M |
3749780 | Certificate of Analysis | 24/01/2024 | DR0801M |
3744986 | Certificate of Analysis | 29/11/2023 | DR0801M |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.