missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LentiArray™ CRISPR Negative Control Lentivirus, human, non-targeting, with GFP

Negative control gRNAs are essential in screening applications for setting thresholds for hit determination.

Brand:  Invitrogen™ A32831

 View more versions of this product

Product Code. 15920383

  • £1612.00 / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

For Research Use Only. Not for use in diagnostic procedures.

This item is not returnable. View return policy

Description

Description

Negative control gRNAs are essential in screening applications for setting thresholds for hit determination.

The Invitrogen™ LentiArray™ CRISPR Negative Control Lentivirus, Human Scrambled, contains a gRNA sequence with no sequence homology to any region of the human genome. This construct also expresses emGFP to provide a visual readout of successful transduction that can be used in determining Multiplicity of Infection (MOI).

The Invitrogen™ LentiArray™ CRISPR Negative Control Lentivirus, Human Scrambled, contains a gRNA sequence with no sequence homology to any region of the human genome. This construct also expresses emGFP to provide a visual readout of successful transduction that can be used in determining Multiplicity of Infection (MOI).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in ultra-cold freezer (-68°C to -85°C).
Tube
gRNA
Genome Editing
gRNA
1 mL
Lentiviral
Negative Control
Human
LentiArray™
U6
Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2685420 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberA32831
Lot Number1880473 Certificate TypeCertificate of Analysis Date19/07/2017 Catalog NumberA32831
Lot Number1831086 Certificate TypeCertificate of Analysis Date31/03/2017 Catalog NumberA32831
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ LentiArray™ CRISPR Negative Control Lentivirus, human, non-targeting, with GFP > 1 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.