missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lipofectamine™ 3000 Transfection Reagent
GREENER_CHOICE

Lipofectamine™ 3000 Transfection Reagent leverages our most advanced lipid nanoparticle technology to provide superior transfection performance with improved application outcomes and reproducible results.

Brand:  Invitrogen™ L3000001

60.83 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.


 View more versions of this product

Product Code. 15282465

  • £91.25 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Animal Origin-Free
Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Plasmid DNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR)
Transfection
Lipofectamine™
0.1 mL
Contains one vial of Lipofectamine™ 3000 Reagent and one vial of P3000TM Enhancer Reagent. Store at 2–8°C. Do not freeze.
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Lipid-based Transfection
Sustainable packaging
Transfection Reagent
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3074156 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberL3000001
Lot Number3035199 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberL3000001
Lot Number2986142 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberL3000001
Lot Number3024515 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberL3000001
Lot Number3074162 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberL3000001
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Lipofectamine™ 3000 Transfection Reagent > 0.1 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.