missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Lab Thermometer
A general purpose/stirring thermometer for a wide application of laboratory needs.
Brand: Fisherbrand™ 63/022/1
Description
- With amber graduations
- Non-roll fitting
- Reinforced bulb for stirring
Specifications
General use | |
155 mm | |
Liquid Filled Thermometer | |
-10°C | |
1°C |
Total Immersion | |
Red Spirit | |
110°C | |
-10°C to +110°C |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 14
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Lab Thermometer > Temperature Range: -10°C to +110°C, Subdivisions: 1°C, Total Immersion, Length: 155 mm
Spot an opportunity for improvement?Share a Content Correction