missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ CELLine™ Bioreactor Flask

Membrane cell culture flasks ideal for antibody and protein production

Brand:  DWK Life Sciences WCL0350-1

238.45 GBP valid until 2025-12-31
Use promo code "24329" to get your promotional price.


 View more versions of this product

Product Code. 12821803

  • £251.00 / Each
Estimated Shipment: 30-06-2025
to see stock.
Explore more special offers
Add to basket

Includes: User manual with feeding and harvesting schedules
This item is not returnable. View return policy

Description

Description

High-density cell culture flasks with dual membrane system allow for less culture maintenance and a high concentration harvest.

  • Bulk media chamber allows for longer run times without media refreshing
  • Cell compartment concentrates cells, antibodies and proteins for simplified purification
  • For suspension cell culture and adherent cell culture: Hybridoma, CHO, NS0, SF, BHK, HEK, etc.

02-912-745, 02-912-742

  • For adherent cell cultivation
  • 1000 mL media compartment
  • 15 mL cell compartment
  • Available as package of one (02-912-745) or package three (02-912-742)
02-912-746, 02-912-743
  • For suspension cell cultivation
  • 350 mL media compartment
  • 5 mL cell compartment
  • Available as package of one (02-912-746) or package five (02-912-746)
02-912-744, 02-912-741
  • For suspension cell cultivation
  • 1000 mL media compartment
  • 15 mL cell compartment
  • Available as package of one (02-912-744) or package three (02-912-741)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.175 oz.
5 mL
Suspension Cultures
1/Cs.
12.3 oz.
350 mL
User manual with feeding and harvesting schedules
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Wheaton™ CELLine™ Bioreactor Flask > Suspension: Media: 350mL; cell: 5mL; pack of 1

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.