Learn More
Falcon™ Cell Culture Multi Flasks



Grow cells faster and easier, increasing productivity in cell culture workflow. Falcon™ Cell Culture Mini Flasks provide even distribution of media across all layers for homogeneous cell growth.
Brand: Falcon™ 353144
152.24 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Increase productivity by enabling to grow more cells faster and easier. 3- or 5-layer formats provide 525 cm2 and 875 cm2 cell growth surface area, respectively.
- Even distribution of media across all layers for homogeneous cell growth
- Ability to mix cells and reagents in the Falcon Multi-Flask saves time
- Flexible design allows to pour or aspirate/recover cells using a pipet
- Consistent surface treatment for predictable scale-up
- Lot number printed on each flask for traceability
- Manufactured in compliance to cGMP standards

Specifications
Polystyrene | |
With cap: 7.8 in. | |
8/Cs. | |
Rectangular | |
5 in. | |
Rectangular | |
875 cm2 |
3.35 in. | |
Straight | |
1/Pk. | |
TC-Treated | |
Sterile | |
Vented |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
5Hours
14Minutes
44Seconds
58
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.