missing translation for 'onlineSavingsMsg'
Learn More
Loratidine, Tocris Bioscience™ Product Code.: 18064292

Loratidine, Tocris Bioscience™

Product Code. 18064292
50 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18064292

Brand: Tocris Bioscience 1944/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Peripheral H1 antagonist; antiallergic agent

Peripheral histamine H1 receptor antagonist (Ki = 35 nM); devoid of central effects. Orally active antiallergic agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79794-75-5
Molecular Formula C22H23ClN2O2
Molecular Weight (g/mol) 382.888
InChI Key JCCNYMKQOSZNPW-UHFFFAOYSA-N
Synonym loratadine, claritin, loratidine, alavert, clarityn, lisino, clarityne, loracert, loradex, bonalerg
PubChem CID 3957
IUPAC Name ethyl 4-(8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate
SMILES CCOC(=O)N1CCC(=C2C3=C(CCC4=C2N=CC=C4)C=C(C=C3)Cl)CC1

Specifications

Quantity 50 mg
Formula Weight 382.88
Percent Purity >99%
Chemical Name or Material Loratidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Loratidine, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.