missing translation for 'onlineSavingsMsg'
Learn More

JZP 361, >99%, Tocris Bioscience™

Potent and selective reversible MAGL inhibitor

Brand:  Tocris Bioscience 5851/10

 View more versions of this product

Product Code. 30000795

  • £90.00 / 10mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Potent and selective reversible MAGL inhibitor (IC50 = 46 nM). Exhibits 35-fold and 150-fold higher selectivity over human FAAH and ABHD6, respectively (IC50 values are 1.79 and 7.24 μM, respectively). Displays affinity for H1 receptors (pA2 = 6.81). Exhibits no activity at cannabinoid receptors at 10 μM.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

[4-(8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)-1-piperidinyl](1H-1,2,4-
>99%
C22H20ClN5O
4-8-chloro-5,6-dihydro-11h-benzo 5,6 cyclohepta 1,2-b pyridin-11-ylidene-1-piperidinyl 1h-1,2,4-triazol-1-yl methanone
GAVZCGTYRWKKDV-UHFFFAOYSA-N
[4-(8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidin-1-yl]-(1,2,4-triazol-1-yl)methanone
102596130
1680193-80-9
C22H20ClN5O
10 mg
Soluble to 100mM in DMSO and to 20mM in ethanol
C1CC2=C(C=CC(=C2)Cl)C(=C3CCN(CC3)C(=O)N4C=NC=N4)C5=C1C=CC=N5
405.886
405.88
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
JZP 361, >99%, Tocris Bioscience™ > 10mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.