Learn More
Invitrogen™ Novex™ Tricine SDS Buffer Kit, includes LC1676 & LC1675
Use with Novex Tricine Gels
Brand:  Invitrogen™ LC1677
99.43 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Product Code. 10758414
  • £168.00 / 1 set
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Includes: Novex Tricine SDS Running Buffer (10X, 500mL - LC1675) and Novex Tricine SDS Sample Buffer (2X, 20mL - LC1676)
Description
Description
In the tricine system developed by Schaegger and von Jagow (1), tricine replaces glycine in the running buffer. This results in more efficient stacking and destacking of low molecular weight proteins and higher resolution of smaller peptides.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Buffer Kit
SDS PAGE Gel
Novex™
The Novex™ Tricine SDS Buffer Kit includes Novex™ Tricine SDS Running Buffer (10X, 500 ml - LC1675) and Novex™ Tricine SDS Sample Buffer (2X, 20 ml - LC1676).
Novex Tricine Gel
1 kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Novex™ Tricine SDS Buffer Kit, includes LC1676 & LC1675 > 1 kit