missing translation for 'onlineSavingsMsg'
Learn More
BRAND™ Low Form Glass Beaker Product Code.: 10738241
In Stock
to see stock.
Add to basket

BRAND™ Low Form Glass Beaker

Product Code. 10738241 Shop All Brand Products
10 mL
Click to view available options
Capacity (Metric):
5 mL
10 mL
50 mL

Product Code. 10738241

Brand: BRAND™ 91212

In Stock
to see stock.
Add to basket
Add to basket

Low Form Glass Beaker features a spout and is constucted of borosilicate glass 3.3.

  • DIN EN ISO 3819
  • Includes a spout
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 10 mL
Material Borosilicate Glass
Includes Spout
Type Low Form
Graduated No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ Low Form Glass Beaker >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.