missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ LPDE Safety Wash Bottles with Leakproof Venting Valve
Shop All Bürkle Products
Click to view available options
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Unlabeled
Description
- Safe filling due to wide-necked opening 39 mm neck diameter
- Permanent print according to GHS with substance name in German, English, French and Spanish, GHS pictograms and CAS number
- LDPE yellow transparent with light protection
- Contents 500 ml
Specifications
Specifications
Capacity (Metric) | 500 mL |
Color | Transparent Yellow |
Material | LDPE |
Closure Color | Red |
Closure Included | Yes |
Product Type | Safety Wash Bottle |
Chemical Label | Ethanol |
Closure Type | Screw Cap |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buerkle™ LPDE Safety Wash Bottles with Leakproof Venting Valve >
Spot an opportunity for improvement?Share a Content Correction