missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lyo-ready Enzymes and Reagents for Recombinase Polymerase Amplification (RPA)

Invitrogen Lyo-ready RPA reagents are designed for fast, sensitive, and specific DNA amplification through isothermal recombinase polymerase amplification (RPA) reactions. Lyo-ready RPA reagents are available in stand-alone and kit formats.

Brand:  Invitrogen™ A72127

 View more versions of this product

Product Code. 30245142

  • £217.00 / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Invitrogen Lyo-ready recombinase polymerase amplification (RPA) reagents are designed for fast, sensitive, and specific DNA amplification through isothermal RPA reactions. Lyo-ready RPA reagents are available in stand-alone and kit formats. Reverse transcriptase can be added to the RPA reaction for amplification of RNA targets. All reagents are glycerol-free, which enables assay lyophilization.

Features of Lyo-ready RPA products

  • Enhanced sensitivity down to 1 copy using optimized (RT-)RPA reaction buffer
  • Robust and specific RNA/DNA amplification
  • Amplification from inhibitor-containing samples
  • Fast target amplification in as little as 20 minutes
  • Ability to easily optimize RPA or RT-RPA reactions

Kit components

The Lyo-ready RPA Kit includes all required components for an RPA reaction:

  • Lyo-ready T4 UvsX Protein—produced from E. coli cells bearing cloned uvsX gene derived from bacteriophage T4
  • Lyo-ready T4 UvsY Protein—a recombination mediator protein that stimulates homologous recombination performed by the UvsX recombinase
  • Lyo-ready Bst DNA Polymerase (Cat. No. A56655)—a proprietary mutant of Bst DNA Polymerase, Large Fragment, with fast reaction speed, increased sensitivity, and tolerance to inhibitors
  • Lyo-ready T4 Gene 32 Protein—a single-stranded DNA binding protein produced from E. coli cells carrying cloned bacteriophage T4 gene 32
  • Optimized reaction buffer, magnesium chloride, and dNTPs
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lyo-ready RPA Kit
RPA, RT-RPA
100 Reactions

• Lyo-ready T4 UvsX Protein (1.2 mg/mL), 50 μL
• Lyo-ready T4 UvsY Protein (1.2 mg/mL), 50 μL
• Lyo-ready Bst DNA Polymerase (6 U/μL), 50 μL
• Lyo-ready T4 Gene 32 Protein (16 mg/mL), 50 μL
• 2X RPA Reaction Buffer, 1000 μL
• 280 mM MgCl2, 100 μL
• 10 mM dNTP Mix, 50 μL

Store at -25°C to -15°C.

SuperScript IV Reverse Transcriptase
End point, real-time, lateral flow
Liquid
Lyo-ready Bst DNA Polymerase
Nucleic Acid Amplification
42°C
Dry Ice
Up to 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3151377 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberA72127
Lot Number3095716 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberA72127
Lot Number3080746 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog NumberA72127
Lot Number3106443 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog NumberA72127
Lot Number3106761 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog NumberA72127
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Lyo-ready Enzymes and Reagents for Recombinase Polymerase Amplification (RPA) > Product: Lyo-ready RPA Kit, Quantity: 100 Reactions, Type: DNA Amplification Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.