missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Lyovapor™ L-200

Freeze dryer with Infinite-Control™

Brand:  BUCHI 11L200IN1422001101

 View more versions of this product

Product Code. 17214724

  • £19055.00 / Each
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High quality freeze drying (-55°C, 6 kg)
  • High level of automation
  • Infinite Control™ includes:
    • Easy method creation
    • Method running
    • Data logging
    • Chart recording in real-time and interruption if needed
Specifications

Specifications

6 kg
4
Stainless Steel
Mobile trolley, Vacuum pump, Drying chamber manifold
23.15 in.
13.2 lb.
-55°C to +30°C
Pfeiffer Duo 6
20.1 in.
240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BUCHI Lyovapor™ L-200 > Lyovapor, Vacuum pump Pfeiffer Duo 6, With mobile trolley, Drying chamber manifold with 4 shelves

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.