Learn More
BUCHI Lyovapor L-200 Shelves 6
Offers high-quality Freeze Drying (-55°C, 6 kg) with a high level of automation. Infinite-Control™ includes easy method creation, method running, data logging, chart recording in real-time and interruption if needed from anywhere at anytime.
Brand:  BUCHI 11L200IN1612001101
 View more versions of this product
Product Code. 17274724
  • £19395.00 / Each
Estimated Shipment: 20-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Compact Lyovapor™ L-200
  • Offers high-quality Freeze Drying (-55 °C, 6 kg) with a high level of automation.
  • Infinite-Control™ includes easy method creation, method running, data logging, chart recording in real-time and interruption if needed from anywhere at anytime.
Specifications
Specifications
6 kg
-55°C to +30°C
No
Advanced vacuum control, No Sample temperature sensor, Trolley
23.1496 in.
6
Stainless Steel
Pfeiffer Duo 6
20.0787 in.
220/240 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Accessories and Services
Services
Show More Show Less
Special Offers
Special Offers
Product Content Correction
Product Title
BUCHI Lyovapor L-200 Shelves 6 > Pfeiffer Duo 6