Learn More
Thermo Scientific™ Lysis Buffer, pH 8.0
For isolation of His-tag modified proteins
Brand: Thermo Scientific™ J63892.AP
45.50 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Colorless | |
Liquid | |
For isolation of His-tag modified proteins | |
Ambient temperatures |
500 mL | |
Cell Lysis Reagent | |
8 |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.