Learn More
Lovibond™ MD 100 3-in-1 Photometer: Chlorine LR, Chlorine HR, Chlorine Dioxide
Provides accurate and reproducible results very quickly. Lovibond MD 100 Photometer uses high quality interference filters with long-life LEDs as a light source without any moving parts in a transparent sample chamber. Other features include ease of operation, ergonomic design, compact dimensions and safe handling.
Brand: Lovibond™ 278000
Includes: Instrument in a carrying case, three round glass vials with lids, one stirring rod and brush, Chlorine E HR (KI) tablets (100 pack), Acidifying GP tablets (100 pack), DPD No. 1 tablets (100 pack), DPD No. 3 tablets (100 pack), Hardcheck P tablets (100 pack), 4 x AAA batteries, warranty information, Certificate of compliance, instruction manual
Description
The calibration and software-based adjustment options mean that the MD 100 is also suitable for use as a testing instrument. The tests are conducted using either Lovibond tablet reagents with long term stability and a guaranteed minimum five or 10 year shelf life, VARIO powder reagents or using liquid reagents.
- Waterproof (as defined in IP68, 1 hour at 0.1m)
- Automatic switch-off
- Real-time clock and date calibration mode
- Backlit LCD display (on keypress)
- Factory calibration and user calibration; Reset to factory calibration possible
- Internal ring memory for 16 data sets
- Infrared interface for test data transfer
Reagents Tablets
Each tablet is hermetically sealed within an individual aluminum foil pocket, protecting against challenging environmental conditions. This packaging keeps each tablet in perfect condition, right up until the time it is needed by the user.
Specifications
- Chlorine LR Range: 0.01 to 6.0mg/L Cl2
- Chlorine HR Range: 5 to 200mg/L Cl2
- Chlorine Dioxide Range: 0.02 to 11mg/L ClO2
- Light source: LED, interference filter (IF) and photosensor in transparent cell chamber
- Wavelength Accuracy: ±1nm
- Photometric Accuracy: 3% FS (T = 20°C to 25°C)
- Photometric Resolution: 0.01 A
- Temperature: 5° to 40°C
- Relative Humidity: 30 to 90% (non condensing)
- Power Supply: 4 AAA batteries
- Operating Time: approx. 17 hours or 5000 test measurements in continuous mode when display backlight is off
- Dimensions (L x W x H): 15.5 x 7.5 x 3.5cm
- Weight: basic unit approx. 260g
Note
Specifications for each type of tablet as well as a Certificate of Analysis for each lot is available in the download area at www.lovibond.com.
Specifications
MD 100 3-in-1 Photometer: Chlorine LR, Chlorine HR, Chlorine Dioxide | |
3% FS | |
Instrument in a carrying case, three round glass vials with lids, one stirring rod and brush, Chlorine E HR (KI) tablets (100 pack), Acidifying GP tablets (100 pack), DPD No. 1 tablets (100 pack), DPD No. 3 tablets (100 pack), Hardcheck P tablets (100 pack), 4 x AAA batteries, warranty information, Certificate of compliance, instruction manual | |
Internal ring memory for 16 data sets | |
0.01 to 6.0mg Cl2, 5 to 200mg/L Cl2, 0.02 to 11mg/L ClO2 | |
260 g |
Backlit LCD | |
3.5 cm | |
15.5 cm | |
LED | |
0.01 A | |
7.5 cm |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- MD100 CHLORINE LR/HR-CHLORINE DIOXIDE TABLETS - PH .
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H372-Causes damage to organs.
- H373-May cause damage to organs.
Precautionary Statement
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P314-Get medical advice/attention if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : potassium iodide, sulphuric acid, citricacid, adipic acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.