Learn More
Thermo Scientific™ Nalgene™ Vented HDPE Closures for Sterile Single Use Erlenmeyer Flasks
Ensure a sterile gas exchange for shaker and suspension cell culture by using Thermo Scientific™ Nalgene™ Vented HDPE Closures for Sterile Single Use Erlenmeyer Flasks.
Brand: Thermo Scientific™ 4114-0038
16.62 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Ensure a sterile gas exchange for shaker and suspension cell culture by using Thermo Scientific™ Nalgene™ Vented HDPE Closures for Sterile Single Use Erlenmeyer Flasks.
- Blue high-density polyethylene with hydrophobic 0.2μm PTFE membrane
- Air exchange without unscrewing closure
- Membrane heat welded into place for effective sealing

Specifications
Blue | |
125, 250mL flasks | |
1 | |
Vented |
HDPE | |
12 | |
Sterile |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.