Learn More
Micronova™ MegaClean™ Heavy-Duty Cleaner
A non-flammable, heavy duty cleaner and degreaser that removes grease, adhesives and photo-resist stains from most surfaces. Available in 1 or 5 gallon bottles and a 1 quart spray bottle.
Brand: Micronova™ MC1-5G
Description
The Micronova MegaClean™ Heavy-Duty Cleaner offers an effective alternative to toxic, flammable, solvent-based cleaners, such as acetone and isopropyl alcohol. Filtered to 0.1 microns, this cleaner is ideal for semiconductor, electronics, aerospace and clean-build operations.
- Non-flammable, heavy duty cleanroom degreaser
- Removes grease, adhesives and photo-resist stains from most surfaces
- Ideal for semiconductor, electronics, aerospace and clean-build operations
- Filtered to 0.1 microns
- Available in 1 or 5 gallon refill bottles and a 1 quart spray bottle
- Biodegradable
- Water soluble
- Effective at low concentrations
Specifications
MegaClean Cleaner | |
Heavy Duty Detergent, Degreaser | |
Mops and Buckets or Wipes and Sponge |
Non-sterile | |
Ideal spot cleaning solution and easily removes grease, adhesives and photo-resist stains from most surfaces, floors, high traffic areas, parts cleaning, adhesive removal | |
5 gal. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
046263-000 | Certificate of Analysis | 08/01/2025 | MC1-5G |
045694-000 | Certificate of Compliance | 15/10/2024 | MC1-5G |
042989-000 | Certificate of Compliance | 17/04/2024 | MC1-5G |
042463-000 | Certificate of Compliance | 26/02/2024 | MC1-5G |
041596-000 | Certificate of Compliance | 14/02/2024 | MC1-5G |