Learn More
Thermo Scientific™ Megafuge ST4 Plus Centrifuge Series
GREENER_CHOICE
Faster processing of more samples with a high capacity-to-footprint ratio and simple programming interface, plus the flexibility to run most tubes and bottles on one centrifuge.
£6034.85 - £15256.65
Specifications
Controller Type Microprocessor
Product Line Megafuge
Display Bright LCD Push-Button User Interface
Drive System Direct, Brushless Induction Low Profile
For Use With (Equipment) Purchase of Thermo Scientific rotor and accessories required for operation
View More Specs
Products 9
Product Code Brand Voltage Rotor Included Model Price Quantity & Availability  
Product Code Brand Voltage Rotor Included Model Price Quantity & Availability  
16380718
GREENER_CHOICE
View accessory products View Documents Promotion Details
Thermo Scientific™
75009503
208/240 V No Megafuge ST4 Plus-MD
£6221.32
1 unit
16330728
GREENER_CHOICE
View accessory products View Documents Promotion Details
Thermo Scientific™
75009518
220/240 V No Megafuge ST4R Plus-MD
£10001.59
1 unit
16390738
GREENER_CHOICE
View accessory products View Documents Promotion Details
Thermo Scientific™
75009903
208/240 V No Megafuge ST4 Plus
£6034.85
1 unit
16340748
GREENER_CHOICE
View accessory products View Documents Promotion Details
Thermo Scientific™
75009918
220/240 V No Megafuge ST4R Plus
£9747.30
1 unit
16300758
View Documents Promotion Details
Thermo Scientific™
75016060
220/240 V Yes Megafuge ST4R Plus-MD, TX-1000 Blood Tube Package
£15256.65
1 unit
16370758
View Documents Promotion Details
Thermo Scientific™
75016069
220/240 V Yes Megafuge ST4R Plus, TX-1000 Cell Culture Package
£14917.62
1 unit
16380758
View Documents Promotion Details
Thermo Scientific™
75016070
208/240 V Yes Megafuge ST4 Plus, TX-1000 Cell Culture Package
£11272.97
1 unit
16370768
View Documents Promotion Details
Thermo Scientific™
75016079
220/240 V Yes Megafuge ST4R Plus-MD, TX-750 Blood Tube Package
£14663.34
1 unit
16310778
View Documents Promotion Details
Thermo Scientific™
75016083
220/240 V Yes Megafuge ST4R Plus, TX-750 Cell Culture Package
£14409.07
1 unit
Description
Show More
Specifications
Specifications
Microprocessor
Bright LCD Push-Button User Interface
Purchase of Thermo Scientific rotor and accessories required for operation
Up to 6 Programs via LCD User Interface buttons
ISO 14971, ISO 13485, ISO 9001
Benchtop Centrifuge
Megafuge
Direct, Brushless Induction Low Profile
Choices of 9 Accel/10 Decel Rates
Up to 9 hr. 59 min.
Option to count up or down
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Megafuge ST4 Plus Centrifuge Series