missing translation for 'onlineSavingsMsg'
Learn More
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals Product Code.: 10163000

Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals

Product Code. 10163000
50 g, Glass bottle
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10163000

Brand: Thermo Scientific Acros 148890500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-51-1
Molecular Formula C2H3NaO2S
Molecular Weight (g/mol) 114.09
MDL Number MFCD00043386
InChI Key GNBVPFITFYNRCN-UHFFFAOYSA-M
Synonym sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoateShow MoreShow Less
PubChem CID 23690444
ChEBI CHEBI:86481
IUPAC Name sodium;2-sulfanylacetate
SMILES C(C(=O)[O-])S.[Na+]

Specifications

Melting Point >300.0°C
Assay Percent Range 97.5% min. (Iodimetry)
Packaging Glass bottle
Linear Formula HSCH2CO2Na
Quantity 50 g
Merck Index 15,8819
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 114.09
Percent Purity 98%
Chemical Name or Material Mercaptoacetic acid, sodium salt, 98%

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460742 Certificate TypeCertificate of Analysis Date01/03/2024 Catalog Number148892500,148890500,148890010
Lot NumberA0458901 Certificate TypeCertificate of Analysis Date22/12/2023 Catalog Number148892500,148890500,148890010
Lot NumberA0453504 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number148892500,148890500,148890010
Lot NumberA0437795 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number148892500,148890500,148890010
Lot NumberA0395660 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number148892500,148890500,148890010
compliance-icons compliance-icons
Product Identifier
  • Mercaptoacetic acid, sodium salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.