missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(Ii) Iodide, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
100 g
500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Quantity | 100 g |
Linear Formula | HgI2 |
UN Number | 1638 |
Solubility Information | Soluble in Potassium Iodide solution |
Formula Weight | 454.4g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Chemical Name or Material | Mercury(Ii) Iodide |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Safety and Handling



Product Identifier
- Mercury(II) iodide
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Mercury diiodide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mercury(Ii) Iodide, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction