missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) sulfate, Honeywell Fluka™

Puriss. p.a., ACS Reagent,≥99.0% (precipitation titration)

£20.94 - £1216.61

Chemical Identifiers

CAS 7783-35-9
Molecular Formula HgO4S
Molecular Weight (g/mol) 296.65
MDL Number MFCD00011047
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
Synonym mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
PubChem CID 24544
IUPAC Name mercury(2+);sulfate
SMILES [Hg++].[O-]S([O-])(=O)=O
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15657280
View Documents Promotion Details
Honeywell-Fluka
83384-10G
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15697280
View Documents Promotion Details
Honeywell-Fluka
83384-50G
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15647280
View Documents Promotion Details
Honeywell-Fluka
83384-100G
100 g
£136.79
100g
Please to purchase this item. Need a web account? Register with us today!
15667280
View Documents Promotion Details
Honeywell-Fluka
83384-250G
250 g
£351.80
250g
Please to purchase this item. Need a web account? Register with us today!
15687280
View Documents Promotion Details
Honeywell-Fluka
83384-500G
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15677280
View Documents Promotion Details
Honeywell-Fluka
83384-3KG
3 kg
£1216.61
3kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7783-35-9
296.65
DOBUSJIVSSJEDA-UHFFFAOYSA-L
24544
[Hg++].[O-]S([O-])(=O)=O
HgO4S
MFCD00011047
mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
mercury(2+);sulfate

Specifications

7783-35-9
HgSO4
UN1645
DOBUSJIVSSJEDA-UHFFFAOYSA-L
mercury(2+);sulfate
24544
≥99% (precipitation titration)
HgO4S
MFCD00011047
mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
[Hg++].[O-]S([O-])(=O)=O
296.65
296.65g/mol
Mercury(II) sulfate
Videos
Safety and Handling

Safety and Handling

P260-P262-P280-P302 + P352 + P310-P304 + P340 + P310

missing translation for 'rtecsNumber' : OX0500000

SDS
Documents

Documents

Product Certifications