missing translation for 'onlineSavingsMsg'
Learn More
Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals Product Code.: 10308160

Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 10308160
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10308160

Brand: Thermo Scientific Acros 210490050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-56-6
Molecular Formula CH5NO·HCl
Molecular Weight (g/mol) 83.52
MDL Number MFCD00012951
InChI Key XNXVOSBNFZWHBV-UHFFFAOYSA-N
Synonym o-methylhydroxylamine hydrochloride, methoxyamine hydrochloride, methoxylamine hydrochloride, methoxyammonium chloride, o-methylhydroxylamine hcl, aminooxy methane hydrochloride, methoxyamine hcl, unii-203546olmf, hydroxylamine, o-methyl-, hydrochloride, o-methylhydroxyamine hydrochlorideShow MoreShow Less
PubChem CID 521874
IUPAC Name O-methylhydroxylamine;hydrochloride
SMILES CON.Cl

Specifications

Melting Point 148°C to 153°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula CH3ONH2·HCl
Quantity 5 g
Beilstein 01, 288
Fieser 01,670
Merck Index 15, 6060
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 83.52
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material Methoxylamine hydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468822 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number210490050,210490250,210491000
Lot NumberA0459254 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number210490050,210490250,210491000
Lot NumberA0459254 Certificate TypeCertificate of Analysis Date16/01/2024 Catalog Number210490050,210490250,210491000
Lot NumberA0448537 Certificate TypeCertificate of Analysis Date20/12/2023 Catalog Number210490050,210490250,210491000
Lot NumberA0425698 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number210490050,210490250,210491000
compliance-icons compliance-icons
Product Identifier
  • Methoxylamine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.