missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Low Conductivity Sensor

Accessory for Mettler Toledo FiveGo portable instruments. Mettler Toledo Low Conductivity Sensors are robust probes from low to medium conductivities (0.01μS/cm to 500μS/cm). LightEdition (LE) sensors are tailor-made for high price/performance ratio.

Brand:  Mettler Toledo™ 51340336

297.55 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


Product Code. 11967841

  • £326.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Ideal for pure water
  • For all common applications
  • Low maintenance and easy handling
  • Includes 1m (39 in.) cable
Note: Requires Mettler Toledo 51302329 LTW-MiniDin Conductivity Adapter. The 51302329 adapter is required for the following meters: FiveGo, Seven2Go, Seven2Go Pro, SevenGo Duo, SevenGo Duo Pro.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.08 cm-1
2 steel poles
38.4 in.
0.01 μS/cm-1
Mini-DIN
Cond probe LE740
1.00 m
Cond probe LE740
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mettler Toledo™ Low Conductivity Sensor > Metal shaft; low conductivity range

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.