Learn More
Cole-Parmer™ Essentials Micro-90 Cleaning Solution
Environmentally friendly cleaning solution, Micro-90 is a mild, yet powerful, free-rinsing cleaner that removes most biological materials. Ideal for use in critical areas and general purpose applications.
Brand: Cole-Parmer™ Essentials 18100-11
277.40 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description
- Removes most biological materials
- Safe for all your ultrasonic cleaners
- Environmentallyl friendly
- Mild but powerful cleaner
- Suitable for critical cleaning and general purpose use
- Radioactive contaminants will not reduce cleaning capability
- Works at any concentration and rinses clean
- Does not contain CFCs, ODCs or borates
Specifications
Liquid | |
NSF-Registered for USDA A1 use | |
Essential Micro-90 cleaning solution | |
CIP, Ultrasonic and Immersion Applications | |
4.22 gal. |
9.5 | |
1:100 to 2:100 | |
Manual or Ultrasonic; Removes Blood and Most Biologoical Materials; Cleans Glass, Most Metals, Plastic, and Rubber | |
4/Cs. | |
4 L |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- MICRO-90 Concentrated Cleaning Solution
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : EDTA Na4
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
50Hours
9Minutes
35Seconds
27New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.