Learn More
Applied Biosystems™ MicroAmp™ Cap Installing Tool
MicroAmp™ Cap Installing Tool
Brand: Applied Biosystems™ 4330015
This item is not returnable.
View return policy
Description
The MicroAmp™ Cap Installing Tool helps you install strips of MicroAmp™ caps onto MicroAmp™ reaction tubes in a tray⁄retainer set or 96-well reaction plate.Press and Seal Caps
The MicroAmp™ Cap Installing Tool enables you to ergonomically seal MicroAmp™ reaction tubes and plates. This tight seal will prevent well-to-well contamination and sample loss.
Used With
MicroAmp™ Optical 8-Cap Strip, MicroAmp™ Optical 8-Tube Strip, 0.2 ml, MicroAmp™ Optical 96-Well Reaction Plate with Barcode, MicroAmp™ Optical 96-Well Reaction Plate with Barcode.
Note: For Research Use Only. Not for use in diagnostics procedures.
Specifications
Cap installing tool for MicroAmp plates, tubes, and caps | |
2720 Thermal Cycler, 7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, GeneAmp 9700, QuantStudio™, SimpliAmp™ Thermal Cycler, StepOne™, StepOnePlus™, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System, ProFlex™ PCR System, ViiA™ 7 Dx Fast System, 7500 Fast Dx System, 2720 Thermal Cycler, ViiA™ 7 Fast System, QuantStudio™ Dx | |
MicroAmp™ | |
1 Tool |
12-cap Strip, 8-cap Strip | |
Sequencing | |
Cap Installing Tool | |
Room Temperature |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Product Title
Applied Biosystems™ MicroAmp™ Cap Installing Tool > 1 tool