Learn More
Carl Zeiss™ AxioCams Microscopy Camera
5 megapixel color microscope camera for documentation in routine labs
Brand:  Carl Zeiss™ 426555-0000-000
 View more versions of this product
Product Code. 15295639
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • CMOS microscope camera, 5 megapixels (2560 x 1920)
  • Sensor size of 1/2.5 in. (diagonal 7.1 mm)
  • Pixel size 2.2 x 2.2 μm
  • Color camera
  • Includes ZEN Lite starter software for image capture and making measurements
Specifications
Specifications
Microscopy Camera AxioCam
AxioCam 105 Color
Axiocam 105 color, driver software 64-bit, USB 3.0 PCIe x1 interface, USB 3.0 connection cable (3 m), SATA power cable and Molex power cable sensor
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Carl Zeiss™ AxioCams Microscopy Camera > Microscopy Camera AxioCam; 105 color