Learn More
SPEX CertiPrep™ HG-400 MiniG™ Tissue Homogenizer and Cell Lyser
The perfect solution for labs looking for a compact yet high-performance tissue homogenizer and cell lyser. Designed to disrupt cellular materials by oscillating deep-well titer plates vertically.
Brand:  SPEX CertiPrep™ 1600
Product Code. 15340422
  • £20680.00 / Each
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Automated mechanical disruption through bead beating,
  • Ideal for high-throughput cell lysis applications including DNA/ RNA extraction, protein extraction and pesticide residue analysis
  • Window in lid allows to view grinding process
  • Lid with safety interlock switch to stops motor if lid is opened during grinding
  • LCD screen displays run times
  • Specifically designed for rapid cell disruption, cell lysis and tissue homogenization through bead beating
(This was formerly the 1600 MiniG Tissue Homogenizer.)
Specifications
Specifications
Tissue Homogenizer
Designed to disrupt cellular materials by oscillating deep-well titer plates vertically
HG-400 MiniG Tissue Homogenizer
1 Ea.
18 kg
1/7 hp
Tissue homogenization, DNA/RNA research and extraction, cell lysis, protein extraction, biofuel research, pesticide residue extraction using QuEChERS or the SweEt method.
500 to 1500 strokes/min.
Animal tissue, plant tissue, cell cultures, fruit, cannabis, seeds, yeast and bacteria.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SPEX CertiPrep™ HG-400 MiniG™ Tissue Homogenizer and Cell Lyser >