missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ (±)-ML 209
RORγt inverse agonist; suppresses Th17 cell differentiation
Brand: Tocris Bioscience™ 5987/10
This item is not returnable.
View return policy
Description
(±)-ML 209 is a RORγt inverse agonist (IC50 = 460 nM). Suppresses Th17 cell differentiation and IL17A expression in vitro. Does not suppress transcription of RORα. Minimal activity displayed on ERα, LXRα and thyroid hormone receptors (IC50 values >4.4 μM).
Specifications
White | |
Nuclear Receptor Antagonists | |
C25H31NO6 | |
Soluble to 100mM in DMSO | |
Inverse agonism Th17 cell differentiation, inhibits MS multiple sclerosis |
Inhibitor | |
1334526-14-5 | |
441g/mol | |
Store at -20°C | |
10 mg |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tocris Bioscience™ (±)-ML 209 > 50mg
For Laboratory Research Use Only
Spot an opportunity for improvement?Share a Content Correction